Anti-MMP21 Rabbit Polyclonal Antibody

ABCAAB11636-50EA 588 EUR
ABCAAB11636-50
Anti-MMP21 Rabbit Polyclonal Antibody
Type: Primary
Antigen: MMP21 (matrix metallopeptidase 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR