Anti-TNF Rat Monoclonal Antibody [clone: MP6-XT3]

Proveedor: Abcam
ABCAAB11564-125EA 0 EUR
ABCAAB11564-125
Anti-TNF Rat Monoclonal Antibody [clone: MP6-XT3]
Anticuerpos
Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: monoclonal
Clone: MP6-XT3
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR