Anti-TNF Rat Monoclonal Antibody PE (Phycoerythrin) [clone: MP6-XT22]

ABCAAB11562-50EA 577 EUR
ABCAAB11562-50
Anti-TNF Rat Monoclonal Antibody PE (Phycoerythrin) [clone: MP6-XT22]
Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: monoclonal
Clone: MP6-XT22
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR