Anti-PVALB Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB11427-50EA 0 EUR
ABCAAB11427-50
Anti-PVALB Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PVALB (parvalbumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR