Anti-Interferon beta Sheep Polyclonal Antibody

ABCAAB11409-50EA 588 EUR
ABCAAB11409-50
Anti-Interferon beta Sheep Polyclonal Antibody
Type: Primary
Antigen: Interferon beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR