Anti-hCG beta 2 epitope Mouse Monoclonal Antibody [clone: INN-hCG-22]

Proveedor: Abcam
ABCAAB11389-250EA 530 EUR
ABCAAB11389-250
Anti-hCG beta 2 epitope Mouse Monoclonal Antibody [clone: INN-hCG-22]
Anticuerpos
Type: Primary
Antigen: hCG beta 2 epitope
Clonality: monoclonal
Clone: INN-hCG-22
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR