Anti-GJB1 Rabbit Polyclonal Antibody

ABCAAB11367-100EA 611 EUR
ABCAAB11367-100
Anti-GJB1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: GJB1 (gap junction protein, beta 1, 32kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR