Anti-HMGB1 Mouse Monoclonal Antibody [clone: 4C3]

Proveedor: Abcam
ABCAAB11354-50EA 770 EUR
ABCAAB11354-50
Anti-HMGB1 Mouse Monoclonal Antibody [clone: 4C3]
Anticuerpos
Type: Primary
Antigen: HMGB1 (high mobility group box 1)
Clonality: monoclonal
Clone: 4C3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR