Anti-Tubulin Mouse Monoclonal Antibody [clone: TUB-1A2]

ABCAAB11325-100EA 611 EUR
ABCAAB11325-100
Anti-Tubulin Mouse Monoclonal Antibody [clone: TUB-1A2]
Type: Primary
Antigen: Tubulin
Clonality: monoclonal
Clone: TUB-1A2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR