Anti-Keratin Guinea pig Polyclonal Antibody

ABCAAB11217-200EA 611 EUR
ABCAAB11217-200
Anti-Keratin Guinea pig Polyclonal Antibody
Type: Primary
Antigen: Keratin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype: none
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR