Anti-Spectrin (alpha + beta) Mouse Monoclonal Antibody [clone: SB-SP1]

ABCAAB11184-100EA 619 EUR
ABCAAB11184-100
Anti-Spectrin (alpha + beta) Mouse Monoclonal Antibody [clone: SB-SP1]
Type: Primary
Antigen: Spectrin (alpha + beta)
Clonality: monoclonal
Clone: SB-SP1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR