Anti-PDYN Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB11137-50EA 806 EUR
ABCAAB11137-50
Anti-PDYN Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PDYN (prodynorphin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR