Anti-APP Mouse Monoclonal Antibody [clone: DE2B4]

Proveedor: Abcam
ABCAAB11132-50EA 0 EUR
ABCAAB11132-50
Anti-APP Mouse Monoclonal Antibody [clone: DE2B4]
Anticuerpos
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone: DE2B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR