Anti-ITGA2B Mouse Monoclonal Antibody [clone: M148]

Proveedor: Abcam
ABCAAB11024-100EA 0 EUR
ABCAAB11024-100
Anti-ITGA2B Mouse Monoclonal Antibody [clone: M148]
Anticuerpos
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone: M148
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR