Anti-MC3R Rabbit Polyclonal Antibody

ABCAAB10986-100EA 604 EUR
ABCAAB10986-100
Anti-MC3R Rabbit Polyclonal Antibody
Type: Primary
Antigen: MC3R (melanocortin 3 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR