Anti-BMP5 Goat Polyclonal Antibody

ABCAAB10858-50EA 671 EUR
ABCAAB10858-50
Anti-BMP5 Goat Polyclonal Antibody
Type: Primary
Antigen: BMP5 (bone morphogenetic protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR