Anti-PDGF BB Goat Polyclonal Antibody

ABCAAB10845-50EA 611 EUR
ABCAAB10845-50
Anti-PDGF BB Goat Polyclonal Antibody
Type: Primary
Antigen: PDGF BB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR