Anti-HIST3H3 Mouse Monoclonal Antibody [clone: mAbcam 10799]

Proveedor: Abcam
ABCAAB10799-100EA 849 EUR
ABCAAB10799-100
Anti-HIST3H3 Mouse Monoclonal Antibody [clone: mAbcam 10799]
Anticuerpos
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: monoclonal
Clone: mAbcam 10799
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR