Anti-IL8 Goat Polyclonal Antibody

ABCAAB10769-50EA 634 EUR
ABCAAB10769-50
Anti-IL8 Goat Polyclonal Antibody
Type: Primary
Antigen: IL8 (interleukin 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR