Anti-IFNAR1 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB10739-25EA 515 EUR
ABCAAB10739-25
Anti-IFNAR1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: IFNAR1 (interferon (alpha, beta and omega) receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR