Anti-WISP1 Rabbit Polyclonal Antibody

ABCAAB10737-100EA 655 EUR
ABCAAB10737-100
Anti-WISP1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: WISP1 (WNT1 inducible signaling pathway protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR