Anti-LIM Kinase 1 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB10561-100EA 753 EUR
ABCAAB10561-100
Anti-LIM Kinase 1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LIM Kinase 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR