Anti-CCND1 Mouse Monoclonal Antibody [clone: DCS-6]

ABCAAB10540-100EA 634 EUR
ABCAAB10540-100
Anti-CCND1 Mouse Monoclonal Antibody [clone: DCS-6]
Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: monoclonal
Clone: DCS-6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR