Anti-BAP1 Mouse Monoclonal Antibody [clone: 3c11]

ABCAAB10531-100EA 577 EUR
ABCAAB10531-100
Anti-BAP1 Mouse Monoclonal Antibody [clone: 3c11]
Type: Primary
Antigen: BAP1 (BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase))
Clonality: monoclonal
Clone: 3c11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR