Anti-c Abl Mouse Monoclonal Antibody [clone: ABL-148]

ABCAAB10528-100EA 611 EUR
ABCAAB10528-100
Anti-c Abl Mouse Monoclonal Antibody [clone: ABL-148]
Type: Primary
Antigen: c Abl
Clonality: monoclonal
Clone: ABL-148
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR