Anti-DCC Goat Polyclonal Antibody

ABCAAB10525-50EA 611 EUR
ABCAAB10525-50
Anti-DCC Goat Polyclonal Antibody
Type: Primary
Antigen: DCC (deleted in colorectal carcinoma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR