Anti-TNFRSF11A Goat Polyclonal Antibody

ABCAAB10497-25EA 611 EUR
ABCAAB10497-25
Anti-TNFRSF11A Goat Polyclonal Antibody
Type: Primary
Antigen: TNFRSF11A (tumor necrosis factor receptor superfamily, member 11a, NFKB activator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR