Anti-HP1 gamma Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB10480-100EA 585 EUR
ABCAAB10480-100
Anti-HP1 gamma Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: HP1 gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human