Anti-CA9 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB10471-100EA 0 EUR
ABCAAB10471-100
Anti-CA9 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR