Anti-CKII alpha Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB10466-50EA 0 EUR
ABCAAB10466-50
Anti-CKII alpha Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CKII alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR