Anti-UCHL1 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB10404-50EA 0 EUR
ABCAAB10404-50
Anti-UCHL1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR