Anti-SMC2 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB10399-50EA 585 EUR
ABCAAB10399-50
Anti-SMC2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SMC2 (structural maintenance of chromosomes 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR