Anti-Macrophage Inflammatory Protein 1 alpha / CCL3 Goat Polyclonal Antibody

ABCAAB10381-100EA 588 EUR
ABCAAB10381-100
Anti-Macrophage Inflammatory Protein 1 alpha / CCL3 Goat Polyclonal Antibody
Type: Primary
Antigen: Macrophage Inflammatory Protein 1 alpha / CCL3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR