Anti-RPA32/RPA2 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB10359-50EA 0 EUR
ABCAAB10359-50
Anti-RPA32/RPA2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: RPA32/RPA2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR