Anti-NPY Guinea pig Polyclonal Antibody

Proveedor: Abcam
ABCAAB10341-50EA 0 EUR
ABCAAB10341-50
Anti-NPY Guinea pig Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NPY (neuropeptide Y)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR