Anti-GRM6 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB10314-50EA 739 EUR
ABCAAB10314-50
Anti-GRM6 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: GRM6 (glutamate receptor, metabotropic 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR