Anti-Mu Opioid Receptor Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB10306-50EA 465 EUR
ABCAAB10306-50
Anti-Mu Opioid Receptor Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Mu Opioid Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR