Anti-GRM2 Rabbit Polyclonal Antibody

ABCAAB10304-100EA 611 EUR
ABCAAB10304-100
Anti-GRM2 Rabbit Polyclonal Antibody
Type: Primary
Antigen: GRM2 (glutamate receptor, metabotropic 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR