Anti-TNPO1 Mouse Monoclonal Antibody [clone: D45]

Proveedor: Abcam
ABCAAB10303-100EA 838 EUR
ABCAAB10303-100
Anti-TNPO1 Mouse Monoclonal Antibody [clone: D45]
Anticuerpos
Type: Primary
Antigen: TNPO1 (transportin 1)
Clonality: monoclonal
Clone: D45
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR