Anti-TNNI3 Mouse Monoclonal Antibody [clone: M155]
Proveedor: Abcam
ABCAAB10237-200EA 550 EUR
ABCAAB10237-200
Anti-TNNI3 Mouse Monoclonal Antibody [clone: M155]
Anticuerpos
Type: Primary
Antigen: TNNI3 (troponin I type 3 (cardiac))
Clonality: monoclonal
Clone: M155
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Dog, Mouse, Rat