Anti-PKA R2 Mouse Monoclonal Antibody [clone: RS30]

ABCAAB10126-500EA 611 EUR
ABCAAB10126-500
Anti-PKA R2 Mouse Monoclonal Antibody [clone: RS30]
Type: Primary
Antigen: PKA R2
Clonality: monoclonal
Clone: RS30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR