Anti-Nicotinic Acetylcholine Receptor alpha 7 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB10096-100EA 790 EUR
ABCAAB10096-100
Anti-Nicotinic Acetylcholine Receptor alpha 7 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Nicotinic Acetylcholine Receptor alpha 7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR