Anti-TNFRSF11B Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9986-100EA 806 EUR
ABCAAB9986-100
Anti-TNFRSF11B Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TNFRSF11B (tumor necrosis factor receptor superfamily, member 11b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR