Anti-CCL7 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9911-100EA 739 EUR
ABCAAB9911-100
Anti-CCL7 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CCL7 (chemokine (C-C motif) ligand 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR