Anti-ALDH1A1 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9883-100EA 833 EUR
ABCAAB9883-100
Anti-ALDH1A1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ALDH1A1 (aldehyde dehydrogenase 1 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR