Anti-TFPI Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9881-100EA 739 EUR
ABCAAB9881-100
Anti-TFPI Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TFPI (tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR