Anti-DEFB4A Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9871-100EA 806 EUR
ABCAAB9871-100
Anti-DEFB4A Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: DEFB4A (defensin, beta 4A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR