Anti-PPP1R15A Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9869-100EA 550 EUR
ABCAAB9869-100
Anti-PPP1R15A Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PPP1R15A (protein phosphatase 1, regulatory subunit 15A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR