Anti-SCGF beta Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9835-100EA 510 EUR
ABCAAB9835-100
Anti-SCGF beta Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SCGF beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR