Anti-Macrophage Inflammatory Protein 3 alpha Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9829-100EA 0 EUR
ABCAAB9829-100
Anti-Macrophage Inflammatory Protein 3 alpha Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Macrophage Inflammatory Protein 3 alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR